missing translation for 'onlineSavingsMsg'
Learn More

5-Benzylamino-1-pentanol 98.0+%, TCI America™

Catalog No. B34615G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B34615G Supplier TCI America Supplier No. B34615G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2937-99-7
Molecular Formula C12H19NO
Molecular Weight (g/mol) 193.29
MDL Number MFCD09688387
InChI Key YMNRQBMCCMQGSH-UHFFFAOYSA-N
Synonym N-(5-Hydroxyamyl)benzylamine, N-(5-Hydroxypentyl)benzylamine
PubChem CID 17206415
IUPAC Name 5-(benzylamino)pentan-1-ol
SMILES C1=CC=C(C=C1)CNCCCCCO

Specifications

Color Yellow
Boiling Point 168°C
Quantity 5 g
Formula Weight 193.29
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 5-Benzylamino-1-pentanol