accessibility menu, dialog, popup

UserName

HNOH

CAS RN 2937-99-7

IUPAC Name: 5-(benzylamino)pentan-1-ol
Synonyms: 5-(benzylamino)pentan-1-ol
Molecular Weight (g/mol): 193.29
Molecular Formula: C12H19NO
InChi Key: YMNRQBMCCMQGSH-UHFFFAOYSA-N
SMILES: OCCCCCNCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results