missing translation for 'onlineSavingsMsg'
Learn More

5-Aminomethyl-2-chloropyridine, 97%

CAS: 97004-04-1 | C6H7ClN2 | 142.586 g/mol

Supplier:  Thermo Scientific Chemicals L1928306

 View more versions of this product

Catalog No. AAL1928306


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

5-Aminomethyl-2-chloropyridine
27°C to 32°C
C6H7ClN2
5 g
8308740
6-chloropyridin-3-yl methanamine, 5-aminomethyl-2-chloropyridine, 2-chloro-5-aminomethylpyridine, 1-6-chloropyridin-3-yl methanamine, 6-chloro-3-picolylamine, 6-chloropyridin-3-yl methylamine, 3-pyridinemethanamine, 6-chloro, 6-chloro-3-pyridyl methylamine, 2-chloro-5-aminomethyl pyridine
C1=CC(=NC=C1CN)Cl
142.586
142.59
97004-04-1
101°C to 102°C (1 mmHg)
MFCD00673153
UN3259
Air Sensitive
XPARFBOWIYMLMY-UHFFFAOYSA-N
(6-chloropyridin-3-yl)methanamine
7020927
97%

Chemical Identifiers

97004-04-1
142.586
XPARFBOWIYMLMY-UHFFFAOYSA-N
7020927
C1=CC(=NC=C1CN)Cl
C6H7ClN2
MFCD00673153
6-chloropyridin-3-yl methanamine, 5-aminomethyl-2-chloropyridine, 2-chloro-5-aminomethylpyridine, 1-6-chloropyridin-3-yl methanamine, 6-chloro-3-picolylamine, 6-chloropyridin-3-yl methylamine, 3-pyridinemethanamine, 6-chloro, 6-chloro-3-pyridyl methylamine, 2-chloro-5-aminomethyl pyridine
(6-chloropyridin-3-yl)methanamine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P261-P264b-P270-P271-P272-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P333+P313-P362-P501c

H301-H315-H317-H318-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only