accessibility menu, dialog, popup

UserName

NNH2Cl

CAS RN 97004-04-1

IUPAC Name: 1-(6-chloropyridin-3-yl)methanamine
Synonyms: 1-(6-chloropyridin-3-yl)methanamine
Molecular Weight (g/mol): 142.59
Molecular Formula: C6H7ClN2
InChi Key: XPARFBOWIYMLMY-UHFFFAOYSA-N
SMILES: NCC1=CN=C(Cl)C=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)