missing translation for 'onlineSavingsMsg'
Learn More

5-Aminomethyl-2-chloropyridine, 97%

Catalog No. p-7047466
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1928303 1 g
AAL1928306 5 g
AAL1928314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1928303 Supplier Thermo Scientific Chemicals Supplier No. L1928303

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 97004-04-1 | C6H7ClN2 | 142.586 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 97004-04-1
Molecular Formula C6H7ClN2
Molecular Weight (g/mol) 142.586
MDL Number MFCD00673153
InChI Key XPARFBOWIYMLMY-UHFFFAOYSA-N
Synonym 6-chloropyridin-3-yl methanamine, 5-aminomethyl-2-chloropyridine, 2-chloro-5-aminomethylpyridine, 1-6-chloropyridin-3-yl methanamine, 6-chloro-3-picolylamine, 6-chloropyridin-3-yl methylamine, 3-pyridinemethanamine, 6-chloro, 6-chloro-3-pyridyl methylamine, 2-chloro-5-aminomethyl pyridine
PubChem CID 7020927
IUPAC Name (6-chloropyridin-3-yl)methanamine
SMILES C1=CC(=NC=C1CN)Cl

Specifications

Melting Point 27°C to 32°C
Boiling Point 101°C to 102°C (1 mmHg)
Quantity 1 g
UN Number UN3259
Beilstein 8308740
Sensitivity Air Sensitive
Formula Weight 142.59
Percent Purity 97%
Chemical Name or Material 5-Aminomethyl-2-chloropyridine
Hazard Category H301-H315-H317-H318-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P333+P313-P362-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only