missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Amino-2-methylbenzonitrile 98.0+%, TCI America™
$32.72 - $96.26
Chemical Identifiers
| CAS | 50670-64-9 |
|---|---|
| Molecular Formula | C8H8N2 |
| Molecular Weight (g/mol) | 132.166 |
| MDL Number | MFCD00017608 |
| InChI Key | YDZVQWCVKXYGIU-UHFFFAOYSA-N |
| Synonym | 5-amino-2-methylbenzenecarbonitrile, 2-methyl-5-aminobenzonitrile, 4-amino-2-cyanotoluene, 3-cyano-4-methylaniline, 5-amino-2-methyl-benzonitrile, benzonitrile, 5-amino-2-methyl, 5-amino-o-tolunitrile, acmc-20a01k, aminomethylbenzenecarbonitrile, 3-amino-6-methylbenzonitrile |
| PubChem CID | 2735365 |
| IUPAC Name | 5-amino-2-methylbenzonitrile |
| SMILES | CC1=C(C=C(C=C1)N)C#N |
Chemical Identifiers
| 50670-64-9 | |
| 132.166 | |
| YDZVQWCVKXYGIU-UHFFFAOYSA-N | |
| 2735365 | |
| CC1=C(C=C(C=C1)N)C#N |
| C8H8N2 | |
| MFCD00017608 | |
| 5-amino-2-methylbenzenecarbonitrile, 2-methyl-5-aminobenzonitrile, 4-amino-2-cyanotoluene, 3-cyano-4-methylaniline, 5-amino-2-methyl-benzonitrile, benzonitrile, 5-amino-2-methyl, 5-amino-o-tolunitrile, acmc-20a01k, aminomethylbenzenecarbonitrile, 3-amino-6-methylbenzonitrile | |
| 5-amino-2-methylbenzonitrile |
Specifications
| 50670-64-9 | |
| 110°C | |
| MFCD00017608 | |
| 3439 | |
| YDZVQWCVKXYGIU-UHFFFAOYSA-N | |
| 5-amino-2-methylbenzonitrile | |
| 2735365 | |
| ≥98.0% (GC,T) | |
| 5-Amino-2-methylbenzonitrile |
| 91°C | |
| C8H8N2 | |
| 1 g | |
| 5-amino-2-methylbenzenecarbonitrile, 2-methyl-5-aminobenzonitrile, 4-amino-2-cyanotoluene, 3-cyano-4-methylaniline, 5-amino-2-methyl-benzonitrile, benzonitrile, 5-amino-2-methyl, 5-amino-o-tolunitrile, acmc-20a01k, aminomethylbenzenecarbonitrile, 3-amino-6-methylbenzonitrile | |
| CC1=C(C=C(C=C1)N)C#N | |
| 132.166 | |
| 132.17 | |
| Crystalline Powder |
Safety and Handling
TSCA : No