missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-methylbenzonitrile 98.0+%, TCI America™

Catalog No. A28141G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A28141G 1 g
A28145G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A28141G Supplier TCI America Supplier No. A28141G

Chemical Identifiers

CAS 50670-64-9
Molecular Formula C8H8N2
Molecular Weight (g/mol) 132.166
MDL Number MFCD00017608
InChI Key YDZVQWCVKXYGIU-UHFFFAOYSA-N
Synonym 5-amino-2-methylbenzenecarbonitrile, 2-methyl-5-aminobenzonitrile, 4-amino-2-cyanotoluene, 3-cyano-4-methylaniline, 5-amino-2-methyl-benzonitrile, benzonitrile, 5-amino-2-methyl, 5-amino-o-tolunitrile, acmc-20a01k, aminomethylbenzenecarbonitrile, 3-amino-6-methylbenzonitrile
PubChem CID 2735365
IUPAC Name 5-amino-2-methylbenzonitrile
SMILES CC1=C(C=C(C=C1)N)C#N

Specifications

Melting Point 91°C
Boiling Point 110°C
Quantity 1 g
UN Number 3439
Formula Weight 132.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-2-methylbenzonitrile