missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-methylbenzonitrile, 97%, Thermo Scientific Chemicals

$133.27 - $397.19

Chemical Identifiers

CAS 50670-64-9
Molecular Formula C8H8N2
Molecular Weight (g/mol) 132.166
MDL Number MFCD00017608
InChI Key YDZVQWCVKXYGIU-UHFFFAOYSA-N
Synonym 5-amino-2-methylbenzenecarbonitrile, 2-methyl-5-aminobenzonitrile, 4-amino-2-cyanotoluene, 3-cyano-4-methylaniline, 5-amino-2-methyl-benzonitrile, benzonitrile, 5-amino-2-methyl, 5-amino-o-tolunitrile, acmc-20a01k, aminomethylbenzenecarbonitrile, 3-amino-6-methylbenzonitrile
PubChem CID 2735365
IUPAC Name 5-amino-2-methylbenzonitrile
SMILES CC1=C(C=C(C=C1)N)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6142206
View Documents
Thermo Scientific Chemicals
H6142206
5 g
Each for $133.27
Only null left
 
AAH6142214
View Documents
Thermo Scientific Chemicals
H6142214
25 g
Each for $397.19
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

50670-64-9
132.166
YDZVQWCVKXYGIU-UHFFFAOYSA-N
2735365
CC1=C(C=C(C=C1)N)C#N
C8H8N2
MFCD00017608
5-amino-2-methylbenzenecarbonitrile, 2-methyl-5-aminobenzonitrile, 4-amino-2-cyanotoluene, 3-cyano-4-methylaniline, 5-amino-2-methyl-benzonitrile, benzonitrile, 5-amino-2-methyl, 5-amino-o-tolunitrile, acmc-20a01k, aminomethylbenzenecarbonitrile, 3-amino-6-methylbenzonitrile
5-amino-2-methylbenzonitrile

Specifications

50670-64-9
C8H8N2
5 g
5-amino-2-methylbenzenecarbonitrile, 2-methyl-5-aminobenzonitrile, 4-amino-2-cyanotoluene, 3-cyano-4-methylaniline, 5-amino-2-methyl-benzonitrile, benzonitrile, 5-amino-2-methyl, 5-amino-o-tolunitrile, acmc-20a01k, aminomethylbenzenecarbonitrile, 3-amino-6-methylbenzonitrile
CC1=C(C=C(C=C1)N)C#N
132.166
132.16
Solid
88°C to 91°C
MFCD00017608
UN3439
YDZVQWCVKXYGIU-UHFFFAOYSA-N
5-amino-2-methylbenzonitrile
2735365
97%
5-Amino-2-methylbenzonitrile
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only