missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Amino-2-methylbenzenesulfonamide 98.0+%, TCI America™
$48.46 - $129.17
Chemical Identifiers
| CAS | 6973-09-7 |
|---|---|
| Molecular Formula | C7H10N2O2S |
| Molecular Weight (g/mol) | 186.23 |
| MDL Number | MFCD06681071 |
| InChI Key | KTPBKMYOIFHJMI-UHFFFAOYSA-N |
| Synonym | 3-amino-6-methylbenzenesulfonamide, 2-methyl-5-aminobenzene sulfonamide, 5-amino-2-methylbenzene-1-sulfonamide, benzenesulfonamide, 5-amino-2-methyl, 5-amino-2-methyl-benzenesulfonamide, 3-aminosulfonyl-4-methylaniline, pubchem14390, p-aminotoluol-sulfonamid, acmc-1bcyh, ksc495k6b |
| PubChem CID | 235511 |
| IUPAC Name | 5-amino-2-methylbenzenesulfonamide |
| SMILES | CC1=C(C=C(C=C1)N)S(=O)(=O)N |
Chemical Identifiers
| 6973-09-7 | |
| 186.23 | |
| KTPBKMYOIFHJMI-UHFFFAOYSA-N | |
| 235511 | |
| CC1=C(C=C(C=C1)N)S(=O)(=O)N |
| C7H10N2O2S | |
| MFCD06681071 | |
| 3-amino-6-methylbenzenesulfonamide, 2-methyl-5-aminobenzene sulfonamide, 5-amino-2-methylbenzene-1-sulfonamide, benzenesulfonamide, 5-amino-2-methyl, 5-amino-2-methyl-benzenesulfonamide, 3-aminosulfonyl-4-methylaniline, pubchem14390, p-aminotoluol-sulfonamid, acmc-1bcyh, ksc495k6b | |
| 5-amino-2-methylbenzenesulfonamide |
Specifications
| 6973-09-7 | |
| C7H10N2O2S | |
| 5 g | |
| KTPBKMYOIFHJMI-UHFFFAOYSA-N | |
| 5-amino-2-methylbenzenesulfonamide | |
| 235511 | |
| ≥98.0% (HPLC,T) | |
| 5-Amino-2-methylbenzenesulfonamide |
| 169°C | |
| MFCD06681071 | |
| 3-amino-6-methylbenzenesulfonamide, 2-methyl-5-aminobenzene sulfonamide, 5-amino-2-methylbenzene-1-sulfonamide, benzenesulfonamide, 5-amino-2-methyl, 5-amino-2-methyl-benzenesulfonamide, 3-aminosulfonyl-4-methylaniline, pubchem14390, p-aminotoluol-sulfonamid, acmc-1bcyh, ksc495k6b | |
| CC1=C(C=C(C=C1)N)S(=O)(=O)N | |
| 186.23 | |
| 186.23 | |
| Crystalline Powder |
Safety and Handling
TSCA : No