missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-methylbenzenesulfonamide, 96%

CAS: 6973-09-7 | C7H10N2O2S | 186.23 g/mol

$52.96 - $180.05

Chemical Identifiers

CAS 6973-09-7
MDL Number MFCD06681071
ChEBI CHEBI:29013
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH3405603
View Documents
Thermo Scientific Chemicals
H3405603
1 g
Each for $52.96
Only null left
 
AAH3405606
View Documents
Thermo Scientific Chemicals
H3405606
5 g
Each for $180.05
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6973-09-7
MFCD06681071
CHEBI:29013

Specifications

6973-09-7
C7H10N2O2S
1 g
KTPBKMYOIFHJMI-UHFFFAOYSA-N
5-amino-2-methylbenzenesulfonamide
235511
96%
163°C to 164°C
MFCD06681071
3-amino-6-methylbenzenesulfonamide, 2-methyl-5-aminobenzene sulfonamide, 5-amino-2-methylbenzene-1-sulfonamide, benzenesulfonamide, 5-amino-2-methyl, 5-amino-2-methyl-benzenesulfonamide, 3-aminosulfonyl-4-methylaniline, pubchem14390, p-aminotoluol-sulfonamid, acmc-1bcyh, ksc495k6b
CC1=C(C=C(C=C1)N)S(=O)(=O)N
186.23
186.23
5-Amino-2-methylbenzenesulfonamide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only