missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-fluorobenzonitrile, 97%

Catalog No. AAH2621006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2621006 5 g
AAH2621014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2621006 Supplier Thermo Scientific Chemicals Supplier No. H2621006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 53312-81-5 | C7H5FN2 | 136.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 53312-81-5
Molecular Formula C7H5FN2
Molecular Weight (g/mol) 136.13
MDL Number MFCD00277872
InChI Key HHTRAISBAAXRKZ-UHFFFAOYSA-N
Synonym 3-cyano-4-fluoroaniline, 2-fluoro-5-aminobenzonitrile, 5-amino-2-fluoro-benzonitrile, benzonitrile, 5-amino-2-fluoro, 4-fluoro-3-cyanoaniline, 3-amino-6-fluorobenzonitrile, ksc494a6l, 3-cyano-4-fluoroanilin, 5-amino-2-fluorobenzenecarbonitrile, buttpark 179\04-11
PubChem CID 2737673
IUPAC Name 5-amino-2-fluorobenzonitrile
SMILES NC1=CC(C#N)=C(F)C=C1

Specifications

Melting Point 92°C to 96°C
Quantity 5 g
UN Number UN3439
Formula Weight 136.13
Percent Purity 97%
Chemical Name or Material 5-Amino-2-fluorobenzonitrile
Hazard Category H315-H318
Hazard Statement GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P362
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only