missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Amino-2-bromobenzotrifluoride 98.0+%, TCI America™
Supplier: TCI America A184725G
Specifications
| 5-Amino-2-bromobenzotrifluoride | |
| 50°C | |
| 108°C | |
| MFCD00007827 | |
| 4-bromo-3-trifluoromethyl aniline, 5-amino-2-bromobenzotrifluoride, 4-bromo-3-trifluoromethyl-phenylamine, 3-trifluoromethyl-4-bromoaniline, 4-amino-1-bromo-2-trifluoromethyl benzene, 4-bromo-alpha,alpha,alpha-trifluoro-m-toluidine, benzenamine, 4-bromo-3-trifluoromethyl, 5-amino-2-bromo benzotrifluoride, buttpark 37\11-68 | |
| NC1=CC=C(Br)C(=C1)C(F)(F)F | |
| 240.02 | |
| 240.02 | |
| Crystalline Powder |
| 393-36-2 | |
| White-Yellow | |
| C7H5BrF3N | |
| 25 g | |
| YGNISOAUPSJDJE-UHFFFAOYSA-N | |
| 4-bromo-3-(trifluoromethyl)aniline | |
| 67852 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 393-36-2 | |
| 240.02 | |
| YGNISOAUPSJDJE-UHFFFAOYSA-N | |
| 67852 | |
| NC1=CC=C(Br)C(=C1)C(F)(F)F |
| C7H5BrF3N | |
| MFCD00007827 | |
| 4-bromo-3-trifluoromethyl aniline, 5-amino-2-bromobenzotrifluoride, 4-bromo-3-trifluoromethyl-phenylamine, 3-trifluoromethyl-4-bromoaniline, 4-amino-1-bromo-2-trifluoromethyl benzene, 4-bromo-alpha,alpha,alpha-trifluoro-m-toluidine, benzenamine, 4-bromo-3-trifluoromethyl, 5-amino-2-bromo benzotrifluoride, buttpark 37\11-68 | |
| 4-bromo-3-(trifluoromethyl)aniline |
Safety and Handling
TSCA : No