missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-bromobenzoic acid, 95%

Catalog No. p-7040576
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6038206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6038206 Supplier Thermo Scientific Chemicals Supplier No. H6038206
Only null left

CAS: 2840-02-0 | C7H6BrNO2 | 216.03 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs and is also used in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs and is also used in medicine.

Solubility
Slightly soluble in water.

Notes
Store at room temperature. Keep away from oxidizing agents.

Chemical Identifiers

CAS 2840-02-0
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.03
MDL Number MFCD07368968
InChI Key FEXDUVBQBNYSQV-UHFFFAOYSA-N
Synonym 5-amino-2-bromo-benzoic acid, 4-bromo-3-carboxyaniline, benzoic acid,5-amino-2-bromo, benzoic acid, 5-amino-2-bromo, pubchem4637, acmc-209h2e, 5-amino-2-bromobenzoicacid, 2-bromo-5-aminobenzoic acid, ksc561g5h, 5-amino-2-bromo benzoic acid
PubChem CID 2769624
IUPAC Name 5-amino-2-bromobenzoic acid
SMILES NC1=CC(C(O)=O)=C(Br)C=C1

Specifications

Melting Point 171°C to 179°C
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 216.03
Percent Purity 95%
Chemical Name or Material 5-Amino-2-bromobenzoic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only