accessibility menu, dialog, popup

UserName

CAS RN 2840-02-0

IUPAC Name: 5-amino-2-bromobenzoic acid
Synonyms: 5-amino-2-bromobenzoic acid
Molecular Weight (g/mol): 216.03
Molecular Formula: C7H6BrNO2
InChi Key: FEXDUVBQBNYSQV-UHFFFAOYSA-N
SMILES: NC1=CC(C(O)=O)=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results