missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-1-naphthol-3-sulfonic Acid Hydrate 97.0+%, TCI America™

Catalog No. A036825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A036825G 25 g
A0368500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A036825G Supplier TCI America Supplier No. A036825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 489-78-1
Molecular Formula C10H9NO4S
Molecular Weight (g/mol) 239.245
MDL Number MFCD00035712
InChI Key GGZZISOUXJHYOY-UHFFFAOYSA-N
Synonym 8-Amino-4-hydroxy-2-naphthalenesulfonic Acid, M Acid
PubChem CID 68106
IUPAC Name 8-amino-4-hydroxynaphthalene-2-sulfonic acid
SMILES C1=CC2=C(C=C(C=C2C(=C1)N)S(=O)(=O)O)O

Specifications

Color Purple-White
Quantity 25 g
Formula Weight 239.25
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-1-naphthol-3-sulfonic Acid Hydrate