missing translation for 'onlineSavingsMsg'
Learn More

5-Acetonyl-2-methoxybenzenesulfonamide 98.0+%, TCI America™

Catalog No. p-7118164
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A29485G 5 g
A294825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A29485G Supplier TCI America Supplier No. A29485G
Only null left

Chemical Identifiers

CAS 116091-63-5
Molecular Formula C10H13NO4S
Molecular Weight (g/mol) 243.28
MDL Number MFCD07782136
InChI Key MQQJFLHZXQRKKJ-UHFFFAOYSA-N
Synonym 2-Methoxy-5-(2-oxopropyl)benzenesulfonamide
PubChem CID 14179759
IUPAC Name 2-methoxy-5-(2-oxopropyl)benzene-1-sulfonamide
SMILES COC1=C(C=C(CC(C)=O)C=C1)S(N)(=O)=O

Specifications

Melting Point 196°C
Quantity 5 g
Formula Weight 243.28
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 5-Acetonyl-2-methoxybenzenesulfonamide