missing translation for 'onlineSavingsMsg'
Learn More

5,8-Dihydroxy-1,4-naphthoquinone 75.0+%, TCI America™

Catalog No. p-7127094
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D20701G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D20701G Supplier TCI America Supplier No. D20701G

Chemical Identifiers

CAS 475-38-7
Molecular Formula C10H6O4
Molecular Weight (g/mol) 190.15
MDL Number MFCD00001685
InChI Key RQNVIKXOOKXAJQ-UHFFFAOYSA-N
Synonym naphthazarin, 5,8-dihydroxy-1,4-naphthoquinone, naphthazarine, naphthazarone, 5,8-dihydroxynaphthoquinone, 5,8-dihydroxy-1,4-naphthalenedione, naphthazalin, 5,8-dihydroxy-1,4-naphthosemiquinone, ccris 6670, dihydroxynaphthoquinone
PubChem CID 10141
ChEBI CHEBI:28849
IUPAC Name 5,8-dihydroxy-1,4-dihydronaphthalene-1,4-dione
SMILES OC1=CC=C(O)C2=C1C(=O)C=CC2=O

Specifications

Melting Point 230°C
Quantity 1 g
Formula Weight 190.15
Percent Purity ≥75.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 5,8-Dihydroxy-1,4-naphthoquinone
RTECSNumber QL7970000
TSCA No