missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ 5,7-Dichlorokynurenic acid sodium salt

Catalog No. 369810R
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
369810R 10 mg
369850R 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 369810R Supplier Tocris Bioscience™ Supplier No. 3698/10*
Only null left

Sodium salt of 5,7-Dichlorokynurenic acid (Cat. No. 0286). Potent NMDA antagonist, acts at glycine site

5,7-Dichlorokynurenic acid sodium salt is a sodium salt of 5,7-Dichlorokynurenic acid (Cat. No. 0286), a potent antagonist at the glycine site of the NMDA receptor (Ki = 79 nM vs. [3H]-glycine).

Specifications

Quantity 10 mg
Product Type 5,7-Dichlorokynurenic acid sodium salt
Purity >98%
Molecular Weight (g/mol) 280.04
Inhibitors 5,7-Dichlorokynurenic acid sodium salt

For Research Use Only