missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dihydro-1,4-dithiin-2,3-dicarboxylic Anhydride 98.0+%, TCI America™

Catalog No. D25755G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D25755G Supplier TCI America Supplier No. D25755G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10489-75-5
Molecular Formula C6H4O3S2
Molecular Weight (g/mol) 188.215
MDL Number MFCD00223990
InChI Key MXSSHXZXAAXCOW-UHFFFAOYSA-N
Synonym 2,3-Dihydro-1,4-dithiino[2,3-c]furan-5,7-dione
PubChem CID 82669
IUPAC Name 2,3-dihydro-[1,4]dithiino[2,3-c]furan-5,7-dione
SMILES C1CSC2=C(S1)C(=O)OC2=O

Specifications

Melting Point 112°C
Quantity 5 g
Formula Weight 188.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5,6-Dihydro-1,4-dithiin-2,3-dicarboxylic Anhydride