accessibility menu, dialog, popup

UserName

CAS RN 10489-75-5

IUPAC Name: 2H,3H,5H,7H-[1,4]dithiino[2,3-c]furan-5,7-dione
Synonyms: 2H,3H-[1,4]dithiino[2,3-c]furan-5,7-dione
Molecular Weight (g/mol): 188.22
Molecular Formula: C6H4O3S2
InChi Key: MXSSHXZXAAXCOW-UHFFFAOYSA-N
SMILES: O=C1OC(=O)C2=C1SCCS2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results