missing translation for 'onlineSavingsMsg'
Learn More

5,6,7,8-Tetrahydro-1-naphthol, 99%

CAS: 529-35-1 | C10H12O | 148.21 g/mol

Supplier:  Thermo Scientific Chemicals L0241618

 View more versions of this product

Catalog No. AAL0241618

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

5,6,7,8-Tetrahydro-1-naphthol
69°C to 71°C
C10H12O
50 g
5,6,7,8-tetrahydro-1-naphthol, 5-hydroxytetralin, 1-naphthalenol, 5,6,7,8-tetrahydro, tetralin-5-ol, 5,6,7,8-tetrahydro-alpha-naphthol, unii-1r7b5i98hv, 5,6,7,8-tetrahydronaphthol, 5,6,7,8-tetrahydro-1-naphthalenol, 5,6,7,8-tetrahydro-naphthalen-1-ol, tetrahydro-.alpha.-naphthol
OC1=CC=CC2=C1CCCC2
148.21
CHEBI:45900
99%
529-35-1
264°C to 265°C (705 mmHg)
MFCD00001734
1865081
SCWNNOCLLOHZIG-UHFFFAOYSA-N
5,6,7,8-tetrahydronaphthalen-1-ol
68258
148.21

Chemical Identifiers

529-35-1
148.21
SCWNNOCLLOHZIG-UHFFFAOYSA-N
68258
5,6,7,8-tetrahydronaphthalen-1-ol
C10H12O
MFCD00001734
5,6,7,8-tetrahydro-1-naphthol, 5-hydroxytetralin, 1-naphthalenol, 5,6,7,8-tetrahydro, tetralin-5-ol, 5,6,7,8-tetrahydro-alpha-naphthol, unii-1r7b5i98hv, 5,6,7,8-tetrahydronaphthol, 5,6,7,8-tetrahydro-1-naphthalenol, 5,6,7,8-tetrahydro-naphthalen-1-ol, tetrahydro-.alpha.-naphthol
CHEBI:45900
OC1=CC=CC2=C1CCCC2
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 208-461-1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only