missing translation for 'onlineSavingsMsg'
Learn More

5,6,7,8-Tetrahydro-1-naphthol, 99%

Catalog No. p-7043737
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0241609 10 g
AAL0241618 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0241609 Supplier Thermo Scientific Chemicals Supplier No. L0241609

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 529-35-1 | C10H12O | 148.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 529-35-1
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.21
MDL Number MFCD00001734
InChI Key SCWNNOCLLOHZIG-UHFFFAOYSA-N
Synonym 5,6,7,8-tetrahydro-1-naphthol, 5-hydroxytetralin, 1-naphthalenol, 5,6,7,8-tetrahydro, tetralin-5-ol, 5,6,7,8-tetrahydro-alpha-naphthol, unii-1r7b5i98hv, 5,6,7,8-tetrahydronaphthol, 5,6,7,8-tetrahydro-1-naphthalenol, 5,6,7,8-tetrahydro-naphthalen-1-ol, tetrahydro-.alpha.-naphthol
PubChem CID 68258
ChEBI CHEBI:45900
IUPAC Name 5,6,7,8-tetrahydronaphthalen-1-ol
SMILES OC1=CC=CC2=C1CCCC2

Specifications

Melting Point 69°C to 71°C
Boiling Point 264°C to 265°C (705 mmHg)
Quantity 10 g
Beilstein 1865081
Formula Weight 148.21
Percent Purity 99%
Chemical Name or Material 5,6,7,8-Tetrahydro-1-naphthol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 208-461-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only