missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5,5'-Thiodisalicylic Acid 98.0+%, TCI America™
$154.02 - $460.11
Chemical Identifiers
| CAS | 1820-99-1 |
|---|---|
| Molecular Formula | C14H10O6S |
| Molecular Weight (g/mol) | 306.288 |
| MDL Number | MFCD00020264 |
| InChI Key | DWMJRSPNFCPIQN-UHFFFAOYSA-N |
| PubChem CID | 74562 |
| IUPAC Name | 5-(3-carboxy-4-hydroxyphenyl)sulfanyl-2-hydroxybenzoic acid |
| SMILES | C1=CC(=C(C=C1SC2=CC(=C(C=C2)O)C(=O)O)C(=O)O)O |
Chemical Identifiers
| 1820-99-1 | |
| 306.288 | |
| DWMJRSPNFCPIQN-UHFFFAOYSA-N | |
| 5-(3-carboxy-4-hydroxyphenyl)sulfanyl-2-hydroxybenzoic acid |
| C14H10O6S | |
| MFCD00020264 | |
| 74562 | |
| C1=CC(=C(C=C1SC2=CC(=C(C=C2)O)C(=O)O)C(=O)O)O |
Specifications
| 1820-99-1 | |
| C14H10O6S | |
| 5 g | |
| C1=CC(=C(C=C1SC2=CC(=C(C=C2)O)C(=O)O)C(=O)O)O | |
| 306.288 | |
| 306.29 | |
| Crystalline Powder |
| White-Yellow | |
| MFCD00020264 | |
| DWMJRSPNFCPIQN-UHFFFAOYSA-N | |
| 5-(3-carboxy-4-hydroxyphenyl)sulfanyl-2-hydroxybenzoic acid | |
| 74562 | |
| ≥98.0% (GC,T) | |
| 5,5′-Thiodisalicylic Acid |
Safety and Handling
TSCA : Yes