missing translation for 'onlineSavingsMsg'
Learn More

5,5'-Methylenedisalicylic acid, 95%

Catalog No. p-163171
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC351730050 5 g Glass bottle
AC351730250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC351730050 Supplier Thermo Scientific Chemicals Supplier No. 351730050

CAS: 122-25-8 | C15H12O6 | 288.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 122-25-8
Molecular Formula C15H12O6
Molecular Weight (g/mol) 288.26
MDL Number MFCD00016506
InChI Key JWQFKVGACKJIAV-UHFFFAOYSA-N
Synonym 5,5'-methylenedisalicylic acid, 5,5'-methylenebis 2-hydroxybenzoic acid, methylenebis salicylic acid, 5,5-methylenebis salicylic acid, unii-2kf4fvv76n, 5,5-methylenedisalicylic acid, 5-3-carboxy-4-hydroxybenzyl salicylic acid, 3,3'-dicarboxy-4,4'-dihydroxydiphenylmethane, 4,4'-dihydroxy-3,3'-dicarboxydiphenylmethane, 2kf4fvv76n
PubChem CID 67145
IUPAC Name 5-[(3-carboxy-4-hydroxyphenyl)methyl]-2-hydroxybenzoic acid
SMILES C1=CC(=C(C=C1CC2=CC(=C(C=C2)O)C(=O)O)C(=O)O)O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 235°C to 242°C
Color Cream
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula CH2[C6H3(OH)COOH]2
Quantity 5 g
Merck Index 15, 6136
Solubility Information Solubility in water: insoluble in water
Formula Weight 288.26
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 5, 5'-Methylenedisalicylic acid, 95%
Show More Show Less
EINECSNumber 204-530-5

RUO – Research Use Only