missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5,5-Dimethoxy-1,2,3,4-tetrachlorocyclopentadiene 95.0+%, TCI America™
Supplier: TCI America T092825G
Specifications
| 5,5-Dimethoxy-1,2,3,4-tetrachlorocyclopentadiene | |
| 27°C | |
| 110°C | |
| MFCD00001353 | |
| 1,2,3,4-Tetrachloro-5,5-dimethoxycyclopentadiene | |
| COC1(C(=C(C(=C1Cl)Cl)Cl)Cl)OC | |
| 263.923 | |
| 263.92 | |
| Crystalline Lumps |
| 2207-27-4 | |
| White-Yellow | |
| C7H6Cl4O2 | |
| 25 g | |
| UHSMEJQTFMHABA-UHFFFAOYSA-N | |
| 1,2,3,4-tetrachloro-5,5-dimethoxycyclopenta-1,3-diene | |
| 75154 | |
| ≥95.0% (GC) |
Chemical Identifiers
| 2207-27-4 | |
| 263.923 | |
| UHSMEJQTFMHABA-UHFFFAOYSA-N | |
| 75154 | |
| COC1(C(=C(C(=C1Cl)Cl)Cl)Cl)OC |
| C7H6Cl4O2 | |
| MFCD00001353 | |
| 1,2,3,4-Tetrachloro-5,5-dimethoxycyclopentadiene | |
| 1,2,3,4-tetrachloro-5,5-dimethoxycyclopenta-1,3-diene |
Safety and Handling
TSCA : No