missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5,5-Dimethoxy-1,2,3,4-tetrachlorocyclopentadiene 95.0+%, TCI America™
$109.74 - $313.80
Chemical Identifiers
| CAS | 2207-27-4 |
|---|---|
| Molecular Formula | C7H6Cl4O2 |
| Molecular Weight (g/mol) | 263.923 |
| MDL Number | MFCD00001353 |
| InChI Key | UHSMEJQTFMHABA-UHFFFAOYSA-N |
| Synonym | 1,2,3,4-Tetrachloro-5,5-dimethoxycyclopentadiene |
| PubChem CID | 75154 |
| IUPAC Name | 1,2,3,4-tetrachloro-5,5-dimethoxycyclopenta-1,3-diene |
| SMILES | COC1(C(=C(C(=C1Cl)Cl)Cl)Cl)OC |
Chemical Identifiers
| 2207-27-4 | |
| 263.923 | |
| UHSMEJQTFMHABA-UHFFFAOYSA-N | |
| 75154 | |
| COC1(C(=C(C(=C1Cl)Cl)Cl)Cl)OC |
| C7H6Cl4O2 | |
| MFCD00001353 | |
| 1,2,3,4-Tetrachloro-5,5-dimethoxycyclopentadiene | |
| 1,2,3,4-tetrachloro-5,5-dimethoxycyclopenta-1,3-diene |
Specifications
| 2207-27-4 | |
| White-Yellow | |
| C7H6Cl4O2 | |
| 5 g | |
| UHSMEJQTFMHABA-UHFFFAOYSA-N | |
| 1,2,3,4-tetrachloro-5,5-dimethoxycyclopenta-1,3-diene | |
| 75154 | |
| ≥95.0% (GC) | |
| 5,5-Dimethoxy-1,2,3,4-tetrachlorocyclopentadiene |
| 27°C | |
| 110°C | |
| MFCD00001353 | |
| 1,2,3,4-Tetrachloro-5,5-dimethoxycyclopentadiene | |
| COC1(C(=C(C(=C1Cl)Cl)Cl)Cl)OC | |
| 263.923 | |
| 263.92 | |
| Crystalline Lumps |
Safety and Handling
TSCA : No