CAS RN 2207-27-4
IUPAC Name:
1,2,3,4-tetrachloro-5,5-dimethoxycyclopenta-1,3-diene
Synonyms:
1,2,3,4-tetrachloro-5,5-dimethoxycyclopenta-1,3-diene
Molecular Weight (g/mol):
263.92
Molecular Formula:
C7H6Cl4O2
InChi Key:
UHSMEJQTFMHABA-UHFFFAOYSA-N
SMILES:
COC1(OC)C(Cl)=C(Cl)C(Cl)=C1Cl
1
–
1
of
1
results
1
5,5-Dimethoxy-1,2,3,4-tetrachlorocyclopentadiene 95.0+%, TCI America™
CAS: 2207-27-4 Molecular Formula: C7H6Cl4O2 Molecular Weight (g/mol): 263.923 MDL Number: MFCD00001353 InChI Key: UHSMEJQTFMHABA-UHFFFAOYSA-N Synonym: 1,2,3,4-Tetrachloro-5,5-dimethoxycyclopentadiene PubChem CID: 75154 IUPAC Name: 1,2,3,4-tetrachloro-5,5-dimethoxycyclopenta-1,3-diene SMILES: COC1(C(=C(C(=C1Cl)Cl)Cl)Cl)OC