missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Bromophenyl)-2-furaldehyde 94.0+%, TCI America™

Catalog No. p-7122857
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32951G 1 g
B32955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B32951G Supplier TCI America Supplier No. B32951G

Chemical Identifiers

CAS 20005-42-9
Molecular Formula C11H7BrO2
Molecular Weight (g/mol) 251.079
MDL Number MFCD00239387
InChI Key QRTAOOSYSVHQGT-UHFFFAOYSA-N
Synonym 5-4-bromophenyl furan-2-carbaldehyde, 5-4-bromophenyl furfural, 5-4-bromophenyl-2-furaldehyde, 5-4-bromophenyl-2-furancarboxaldehyde, 5-4-bromophenyl-2-furancarbaldehyde, 2-furancarboxaldehyde, 5-4-bromophenyl, 5-4-bromo-phenyl-furan-2-carbaldehyde, 5-p-bromophenyl furancarboxaldehyde, 5-p-bromophenyl furfural, 5-4-bromo-phenyl-furfural
PubChem CID 619577
IUPAC Name 5-(4-bromophenyl)furan-2-carbaldehyde
SMILES C1=CC(=CC=C1C2=CC=C(O2)C=O)Br

Specifications

Melting Point 153°C
Quantity 1 g
Formula Weight 251.08
Percent Purity ≥94.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-(4-Bromophenyl)-2-furaldehyde