accessibility menu, dialog, popup

UserName

BrOO

CAS RN 20005-42-9

IUPAC Name: 5-(4-bromophenyl)furan-2-carbaldehyde
Synonyms: 5-(4-bromophenyl)furan-2-carbaldehyde
Molecular Weight (g/mol): 251.08
Molecular Formula: C11H7BrO2
InChi Key: QRTAOOSYSVHQGT-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)C1=CC=C(O1)C=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results