missing translation for 'onlineSavingsMsg'
Learn More

5-[4-(3,5-Dimethyl-4-nitrostyryl)benzamido]-2-(6-hydroxy-3-oxo-3H-xanthene-9-yl)benzoic Acid Monohydrate 90.0+%, TCI America™

Catalog No. d39595mg
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D39595MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D39595MG Supplier TCI America Supplier No. D39595MG

Chemical Identifiers

CAS 1160555-05-4
Molecular Formula C37H26N2O8
Molecular Weight (g/mol) 626.621
InChI Key UQQWUXKLZVVNDR-ONEGZZNKSA-N
Synonym 4-[4-(3,5-Dimethyl-4-nitrostyryl)benzamido]fluorescein, Flu-DNB
PubChem CID 101858728
IUPAC Name 5-[[4-[(E)-2-(3,5-dimethyl-4-nitrophenyl)ethenyl]benzoyl]amino]-2-(3-hydroxy-6-oxoxanthen-9-yl)benzoic acid
SMILES CC1=CC(=CC(=C1[N+](=O)[O-])C)C=CC2=CC=C(C=C2)C(=O)NC3=CC(=C(C=C3)C4=C5C=CC(=O)C=C5OC6=C4C=CC(=C6)O)C(=O)O

Specifications

Melting Point 300°C
Color Yellow
Quantity 5 mg
Formula Weight 626.62
Percent Purity ≥90.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 5-[4-(3,5-Dimethyl-4-nitrostyryl)benzamido]-2-(6-hydroxy-3-oxo-3H-xanthene-9-yl)benzoic Acid Monohyd
TSCA No
Recommended Storage Freezer