accessibility menu, dialog, popup

UserName

CAS RN 1160555-05-4

IUPAC Name: 5-{4-[2-(3,5-dimethyl-4-nitrophenyl)ethenyl]benzamido}-2-(6-hydroxy-3-oxo-3H-xanthen-9-yl)benzoic acid
Synonyms: 5-{4-[2-(3,5-dimethyl-4-nitrophenyl)ethenyl]benzamido}-2-(3-hydroxy-6-oxoxanthen-9-yl)benzoic acid
Molecular Weight (g/mol): 626.62
Molecular Formula: C37H26N2O8
InChi Key: UQQWUXKLZVVNDR-UHFFFAOYSA-N
SMILES: CC1=CC(C=CC2=CC=C(C=C2)C(=O)NC2=CC(C(O)=O)=C(C=C2)C2=C3C=CC(=O)C=C3OC3=CC(O)=CC=C23)=CC(C)=C1[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results