missing translation for 'onlineSavingsMsg'
Learn More

(4S)-(-)-4-Isopropyl-5,5-diphenyl-2-oxazolidinone 98.0+%, TCI America™

Catalog No. I07621G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I07621G 1 g
I07625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I07621G Supplier TCI America Supplier No. I07621G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 184346-45-0
Molecular Formula C18H19NO2
Molecular Weight (g/mol) 281.355
MDL Number MFCD03093556
InChI Key PHTOJBANGYSTOH-INIZCTEOSA-N
PubChem CID 9838646
IUPAC Name (4S)-5,5-diphenyl-4-propan-2-yl-1,3-oxazolidin-2-one
SMILES CC(C)C1C(OC(=O)N1)(C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 254°C
Color White
Quantity 1 g
Formula Weight 281.36
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material (4S)-(-)-4-Isopropyl-5,5-diphenyl-2-oxazolidinone