accessibility menu, dialog, popup

UserName

CAS RN 184346-45-0

IUPAC Name: 5,5-diphenyl-4-(propan-2-yl)-1,3-oxazolidin-2-one
Synonyms: 4-isopropyl-5,5-diphenyl-1,3-oxazolidin-2-one
Molecular Weight (g/mol): 281.36
Molecular Formula: C18H19NO2
InChi Key: PHTOJBANGYSTOH-UHFFFAOYNA-N
SMILES: CC(C)C1NC(=O)OC1(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results