missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Labs Spectra/Por™ 6 1000 D MWCO Standard RC Pre-wetted Dialysis Kits

With Pre-wetted ready-to-use tubing, not requiring soaking to remove glycerol

$371.82 - $371.82

Specifications

MWCO 1 kDa
Quantity 1 m Tubing/Kit
View More Specs

Includes:
  • 1 meter of Spectra/Por 6 Dialysis Tubing
  • 1 Standard Spectra/Por Closure (for buoyancy)
  • 1 Weighted Spectra/Por Closure (for orientation)
  • 5 Tubing Opening Picks

Products 3
Catalog Number Mfr. No. Diameter (Metric) Length (Metric) Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Length (Metric) Price Quantity  
08-700-196
View Documents
Spectrum™
132638T
24 mm 4.6 mL/cm vol./length
Each for $371.82
Only null left
 
08-700-195
View Documents
Spectrum™
132636T
11.5 mm 1.1 mL/cm vol./length
Each for $371.82
Only null left
 
08-700-197
View Documents
Spectrum™
132640T
29 mm 6.4 mL/cm vol./length
Each for $371.82
Only null left
 
Description

Description

  • Clear, flexible, study standard grade Regenerated Cellulose (RC) dialysis membrane
  • Membrane packaged wet in preservative solution of 0.05% sodium azide
  • Kits available in 1m tubing lengths ideal for membrane evaluation and qualification
  • Include a pair of closures and convenient tubing opening picks

Specifications

Specifications

1 kDa
1 m Tubing/Kit
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'