missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum™ Labs Spectra/Por™ 6 1000 D MWCO Standard RC Pre-wetted Dialysis Kits
Description
Includes
- 1 meter of Spectra/Por 6 Dialysis Tubing
- 1 Standard Spectra/Por Closure (for buoyancy)
- 1 Weighted Spectra/Por Closure (for orientation)
- 5 Tubing Opening Picks
- Clear, flexible, study standard grade Regenerated Cellulose (RC) dialysis membrane
- Membrane packaged wet in preservative solution of 0.05% sodium azide
- Kits available in 1m tubing lengths ideal for membrane evaluation and qualification
- Include a pair of closures and convenient tubing opening picks
Specifications
Specifications
| Product Type | Dialysis Trial Kit |
| MWCO | 1 kDa |
| Format | Tubing |
| Diameter (Metric) | 24 mm |
| Length (Metric) | 4.6 mL/cm vol./length |
| Quantity | 1 m Tubing/Kit |