missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Biohazard Identification Signs: Biohazard Infectious Waste
For identification of equipment, rooms and materials that contain, or are contaminated with, hazardous agents
$20.40 - $37.86
Specifications
| Height (English) | 14 in. |
|---|---|
| Height (Metric) | 4.2 m |
| Width (English) | 10 in. |
| Width (Metric) | 3 m |
| Displayed Text | BIOHAZARD INFECTIOUS WASTE |
Description
- Made from aluminum, plastic or self-sticking polyester
- Size: 14 in. H x 10 in. W
- Legend: BIOHAZARD INFECTIOUS WASTE
Specifications
| 14 in. | |
| 10 in. | |
| BIOHAZARD INFECTIOUS WASTE |
| 4.2 m | |
| 3 m | |
| Biohazard Identification Sign |