missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Biohazard Identification Signs: Biohazard Infectious Waste

Catalog No. 19140986
Change view
Click to view available options
Material:
Aluminum
Plastic
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19140986 Plastic
19140992 Aluminum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19140986 Supplier Brady™ Supplier No. 126640

For identification of equipment, rooms and materials that contain, or are contaminated with, hazardous agents

  • Made from aluminum, plastic or self-sticking polyester
  • Size: 14 in. H x 10 in. W
  • Legend: BIOHAZARD INFECTIOUS WASTE

Specifications

Material Plastic
Height (English) 14 in.
Height (Metric) 4.2 m
Width (English) 10 in.
Width (Metric) 3 m
Displayed Text BIOHAZARD INFECTIOUS WASTE
Product Type Biohazard Identification Sign