missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Biohazard Identification Signs: Biohazard Infectious Waste

Catalog No. 19140992
Change view
Click to view available options
Material:
Aluminum
Plastic
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19140992 Aluminum
19140986 Plastic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19140992 Supplier Brady™ Supplier No. 126639

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For identification of equipment, rooms and materials that contain, or are contaminated with, hazardous agents

  • Made from aluminum, plastic or self-sticking polyester
  • Size: 14 in. H x 10 in. W
  • Legend: BIOHAZARD INFECTIOUS WASTE

Specifications

Material Aluminum
Mounting Type Corner Holes
Height (English) 14 in.
Height (Metric) 4.2 m
Width (English) 10 in.
Width (Metric) 3 m
Shape Rectangular
Languages English
Displayed Text BIOHAZARD INFECTIOUS WASTE
Product Type Biohazard Identification Sign
Color Black on Orange
Show More Show Less