missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Bench Riser Kit

The pre-configured components of Benchpro Bench Riser Kit make lab setup simple.

$1539.00 - $1900.87

Specifications

Includes Steel drawer 15.2cm (6 in.), 4 outlets frame power, 8 outlets power strip, riser shelf
Product Type Bench Riser Kit
View More Specs

Includes: Steel drawer 15.2cm (6 in.), 4 outlets frame power, 8 outlets power strip, riser shelf

Products 3
Catalog Number Mfr. No. Height (English) Price Quantity  
Catalog Number Mfr. No. Height (English) Price Quantity  
25-100-328
View Documents
Fisherbrand™
72FSRKIT
72 in.
Each for $1,900.87
Only null left
 
25-100-326
View Documents
Fisherbrand™
48FSRKIT
48 in.
Each for $1,539.00
Only null left
 
25-100-327
View Documents
Fisherbrand™
60FSRKIT
60 in.
Each for $1,691.15
Only null left
 
Description

Description

The pre-configured components of Bench Riser Kit make lab setup simple. FisherBrand Riser Kit include steel drawer, built-in power frame, power strip, riser Shelf. White Steel. 25 year unconditional manufacturer warranty

  • Pre-configured for simple setup
  • Steel drawer
  • Built-in power frame
  • Power strip
  • Riser shelf
  • White steel
  • 25 year unconditional warranty

Order Info

Kit does NOT include the workbench/table

Warranty

25-year manufacturer warranty

Specifications

Specifications

Steel drawer 15.2cm (6 in.), 4 outlets frame power, 8 outlets power strip, riser shelf
Bench Riser Kit
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'