Learn More
Fisherbrand™ Bench Riser Kit
Description
Includes
Kit include steel drawer, built-in power frame, power strip, riser shelf
The pre-configured components of Bench Riser Kit make lab setup simple. FisherBrand Riser Kit include steel drawer, built-in power frame, power strip, riser Shelf. White Steel. 25 year unconditional manufacturer warranty
- Pre-configured for simple setup
- Steel drawer
- Built-in power frame
- Power strip
- Riser shelf
- White steel
- 25 year unconditional warranty
Order Info
Kit does NOT include the workbench/table
Warranty
25-year manufacturer warranty
Specifications
Specifications
| Product Type | Bench Riser Kit |
| Includes | Steel drawer 15.2cm (6 in.), 4 outlets frame power, 8 outlets power strip, riser shelf |
| Height (English) | 72 in. |
| Height (Metric) | 182.8 cm |