missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Native pepsin from Porcine Stomach Mucosa

Catalyzes hydrolysis of aminoacyl-proline to amino acid and proline

$78.60 - $346.35

Specifications

Molecular Weight (g/mol) 35,000g/mole
Name Native pepsin
pH Range 2 - 4
Source Porcine stomach mucosa
Storage Requirements -20°C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
51-636-0500MG
View Documents
MilliporeSigma™
516360500MG
500 mg
Each for $78.60
Only null left
 
51-636-025GM
View Documents
MilliporeSigma™
51636025GM
2.5 g
Each for $346.35
Only null left
 
Description

Description

  • Secreted by mucosal lining of stomach
  • Active under acidic conditions
  • Inhibitors include aliphatic alcohols, pepstatin A, and pH: >6.0
  • Lyophilized
  • Molar mass: 35,000g/mole
  • pH: 2 - 4

Specifications

Specifications

35,000g/mole
2 - 4
-20°C
Native pepsin
Porcine stomach mucosa
Videos
SDS
Documents

Documents