missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Native pepsin from Porcine Stomach Mucosa
Catalyzes hydrolysis of aminoacyl-proline to amino acid and proline
Supplier: MilliporeSigma™ 51636025GM
Description
- Secreted by mucosal lining of stomach
- Active under acidic conditions
- Inhibitors include aliphatic alcohols, pepstatin A, and pH: >6.0
- Lyophilized
- Molar mass: 35,000g/mole
- pH: 2 - 4
Specifications
| 35,000g/mole | |
| 2 - 4 | |
| Porcine stomach mucosa |
| Native pepsin | |
| 2.5 g | |
| -20°C |