missing translation for 'onlineSavingsMsg'
Learn More

4-Vinylpyridine, 95%, stab. with 80 to 150ppm hydroquinone

CAS: 100-43-6 | C7H7N | 105.14 g/mol

Supplier:  Thermo Scientific Chemicals L13316AE

 View more versions of this product

Catalog No. AAL13316AE


Only null left
Explore available promotions

Description

Description

4-Vinylpyridine acts as a co-monomer in styrene-butadiene polymers to promote adhesion between the rubber compound and supporting fibers (or cords) in tires, belts and hoses. It serves as an intermediate for manufacturing chemicals and pharmaceuticals. Further, it is used as a reagent for the modification of cysteine. In addition to this, it is used to make poly(4-vinylpyridine) and elastomers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Vinylpyridine acts as a co-monomer in styrene-butadiene polymers to promote adhesion between the rubber compound and supporting fibers (or cords) in tires, belts and hoses. It serves as an intermediate for manufacturing chemicals and pharmaceuticals. Further, it is used as a reagent for the modification of cysteine. In addition to this, it is used to make poly(4-vinylpyridine) and elastomers.

Solubility
Miscible with water, chloroform, ethanol, dilute acids, alcohol, hydrocarbons, esters and ketones.

Notes
Light sensitive. Incompatible with strong acids, strong oxidizing agents, ultraviolet radiation and free radical initiators.
Specifications

Specifications

4-Vinylpyridine
100-43-6
58°C to 61°C (12 mmHg)
C7H7N
MFCD00006447
UN3073
4-vinylpyridine, pyridine, 4-ethenyl, pyridine, 4-vinyl, gamma-vinylpyridine, unii-i56g67xm8d, ccris 5240, 4vp, 4-pyridylethylene, 4-vinyl-pyridine, 4-vinyl pyridine
KFDVPJUYSDEJTH-UHFFFAOYSA-N
4-ethenylpyridine
7502
96%
stabilized with 100ppm hydroquinone
0.984
51°C (124°F)
1.551
100 mL
104506
Miscible with water,chloroform,ethanol,dilute acids,alcohol,hydrocarbons,esters and ketones.
C=CC1=CC=NC=C1
105.14
105.14

Chemical Identifiers

100-43-6
105.14
KFDVPJUYSDEJTH-UHFFFAOYSA-N
7502
C=CC1=CC=NC=C1
C7H7N
MFCD00006447
4-vinylpyridine, pyridine, 4-ethenyl, pyridine, 4-vinyl, gamma-vinylpyridine, unii-i56g67xm8d, ccris 5240, 4vp, 4-pyridylethylene, 4-vinyl-pyridine, 4-vinyl pyridine
4-ethenylpyridine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331-H334-H314-H318-H226-H317
Toxic if swallowed.
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
May cause an allergic skin reaction.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P272-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c

H226-H301+H311+H331-H314-H317-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: VINYLPYRIDINES, STABILIZED

EINECSNumber : 202-852-0

RTECSNumber : UU1045000

TSCA : Yes

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only