missing translation for 'onlineSavingsMsg'
Learn More

4-Vinylpyridine, 95%, stab. with 80 to 150ppm hydroquinone

Catalog No. p-7046152
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL13316AC 25 mL
AAL13316AE 100 mL
AAL13316AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL13316AC Supplier Thermo Scientific Chemicals Supplier No. L13316AC
Only null left

CAS: 100-43-6 | C7H7N | 105.14 g/mol

4-Vinylpyridine acts as a co-monomer in styrene-butadiene polymers to promote adhesion between the rubber compound and supporting fibers (or cords) in tires, belts and hoses. It serves as an intermediate for manufacturing chemicals and pharmaceuticals. Further, it is used as a reagent for the modification of cysteine. In addition to this, it is used to make poly(4-vinylpyridine) and elastomers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Vinylpyridine acts as a co-monomer in styrene-butadiene polymers to promote adhesion between the rubber compound and supporting fibers (or cords) in tires, belts and hoses. It serves as an intermediate for manufacturing chemicals and pharmaceuticals. Further, it is used as a reagent for the modification of cysteine. In addition to this, it is used to make poly(4-vinylpyridine) and elastomers.

Solubility
Miscible with water, chloroform, ethanol, dilute acids, alcohol, hydrocarbons, esters and ketones.

Notes
Light sensitive. Incompatible with strong acids, strong oxidizing agents, ultraviolet radiation and free radical initiators.

Chemical Identifiers

CAS 100-43-6
Molecular Formula C7H7N
Molecular Weight (g/mol) 105.14
MDL Number MFCD00006447
InChI Key KFDVPJUYSDEJTH-UHFFFAOYSA-N
Synonym 4-vinylpyridine, pyridine, 4-ethenyl, pyridine, 4-vinyl, gamma-vinylpyridine, unii-i56g67xm8d, ccris 5240, 4vp, 4-pyridylethylene, 4-vinyl-pyridine, 4-vinyl pyridine
PubChem CID 7502
IUPAC Name 4-ethenylpyridine
SMILES C=CC1=CC=NC=C1

Specifications

Density 0.984
Boiling Point 58°C to 61°C (12 mmHg)
Flash Point 51°C (124°F)
Refractive Index 1.551
Quantity 25 mL
UN Number UN3073
Beilstein 104506
Solubility Information Miscible with water,chloroform,ethanol,dilute acids,alcohol,hydrocarbons,esters and ketones.
Formula Weight 105.14
Percent Purity 96%
Chemical Name or Material 4-Vinylpyridine, stabilized with 100ppm hydroquinone
Show More Show Less
Hazard Category H226-H301+H311+H331-H314-H317-H335
Hazard Statement GHS H Statement
H301-H331-H334-H314-H318-H226-H317
Toxic if swallowed.
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
May cause an allergic skin reaction.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P272-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: VINYLPYRIDINES, STABILIZED
EINECSNumber 202-852-0
RTECSNumber UU1045000
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only