accessibility menu, dialog, popup

UserName

H2CN

CAS RN 100-43-6

IUPAC Name: 4-ethenylpyridine
Synonyms: 4-vinylpyridine
Molecular Weight (g/mol): 105.14
Molecular Formula: C7H7N
InChi Key: KFDVPJUYSDEJTH-UHFFFAOYSA-N
SMILES: C=CC1=CC=NC=C1
Boiling Point 
  • (3)
  • (3)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (6)
Physical Form 
  • (11)
Quantity 
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (2)
  • (9)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (6)
  • (11)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)