missing translation for 'onlineSavingsMsg'
Learn More

4-Vinylbenzoic Acid (stabilized with BHT) 97.0+%, TCI America™

Catalog No. V00591G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V00591G 1 g
V00595G 5 g
V005925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. V00591G Supplier TCI America Supplier No. V00591G

Chemical Identifiers

CAS 1075-49-6
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.161
MDL Number MFCD00002569
InChI Key IRQWEODKXLDORP-UHFFFAOYSA-N
Synonym 4-vinylbenzoic acid, p-vinylbenzoic acid, 4-carboxystyrene, benzoic acid, 4-ethenyl, styrene-4-carboxylic acid, 4-vinyl-benzoic acid, p-vinyl benzoic acid, 4-vinyl benzoic acid, para-vinyl benzoic acid, pubchem12624
PubChem CID 14098
IUPAC Name 4-ethenylbenzoic acid
SMILES C=CC1=CC=C(C=C1)C(=O)O

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 148.16
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Vinylbenzoic Acid (stabilized with BHT)
TSCA No
Recommended Storage Refrigerator