accessibility menu, dialog, popup

UserName

CAS RN 1075-49-6

IUPAC Name: 4-ethenylbenzoic acid
Synonyms: benzoic acid, 4-ethenyl-
Molecular Weight (g/mol): 148.16
Molecular Formula: C9H8O2
InChi Key: IRQWEODKXLDORP-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=C)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (6)
Percent Purity 
  • (3)
  • (1)
  • (1)
  • (3)
Quantity 
  • (3)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (2)
  • (6)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)