missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Ureidobenzeneboronic acid pinacol ester, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H5304603
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| 4-Ureidobenzeneboronic acid pinacol ester | |
| 124°C to 128°C | |
| MFCD06795683 | |
| 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl urea, 4-ureido phenylboronic acid, pinacol ester, 4-carbamoylamino benzeneboronic acid, pinacol ester, 4-ureidophenylboronic acid pinacol ester, n-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl urea, 4-n-aminocarbonyl aminophenylboronic acid, pinacol ester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl phenylurea, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl urea, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenylurea, 4-ureidobenzeneboronic acid pinacol ester | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)NC(=O)N | |
| 262.116 | |
| 262.12 |
| 877134-77-5 | |
| C13H19BN2O3 | |
| 1 g | |
| BVWCMMVUJQHJJE-UHFFFAOYSA-N | |
| [4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]urea | |
| 16727440 | |
| 98% |
Chemical Identifiers
| 877134-77-5 | |
| 262.116 | |
| BVWCMMVUJQHJJE-UHFFFAOYSA-N | |
| 16727440 | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)NC(=O)N |
| C13H19BN2O3 | |
| MFCD06795683 | |
| 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl urea, 4-ureido phenylboronic acid, pinacol ester, 4-carbamoylamino benzeneboronic acid, pinacol ester, 4-ureidophenylboronic acid pinacol ester, n-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl urea, 4-n-aminocarbonyl aminophenylboronic acid, pinacol ester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl phenylurea, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl urea, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenylurea, 4-ureidobenzeneboronic acid pinacol ester | |
| [4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]urea |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures