missing translation for 'onlineSavingsMsg'
Learn More

4-Ureidobenzeneboronic acid pinacol ester, 98%, Thermo Scientific™

Catalog No. p-7035331
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH53046MD 250 mg
AAH5304603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH53046MD Supplier Thermo Scientific Chemicals Supplier No. H53046MD
Only null left

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 877134-77-5
Molecular Formula C13H19BN2O3
Molecular Weight (g/mol) 262.116
MDL Number MFCD06795683
InChI Key BVWCMMVUJQHJJE-UHFFFAOYSA-N
Synonym 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl urea, 4-ureido phenylboronic acid, pinacol ester, 4-carbamoylamino benzeneboronic acid, pinacol ester, 4-ureidophenylboronic acid pinacol ester, n-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl urea, 4-n-aminocarbonyl aminophenylboronic acid, pinacol ester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl phenylurea, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl urea, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenylurea, 4-ureidobenzeneboronic acid pinacol ester
PubChem CID 16727440
IUPAC Name [4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]urea
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)NC(=O)N

Specifications

Melting Point 124°C to 128°C
Quantity 250 mg
Formula Weight 262.12
Percent Purity 98%
Chemical Name or Material 4-Ureidobenzeneboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures