missing translation for 'onlineSavingsMsg'
Learn More

4-Tritylphenol, 98%

Catalog No. AAL0607306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0607306 5 g
AAL0607314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0607306 Supplier Thermo Scientific Chemicals Supplier No. L0607306

CAS: 978-86-9 | C25H20O | 336.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 978-86-9
Molecular Formula C25H20O
Molecular Weight (g/mol) 336.43
MDL Number MFCD00002364
InChI Key NIPKXTKKYSKEON-UHFFFAOYSA-N
Synonym 4-triphenylmethylphenol, 4-triphenylmethyl phenol, p-tritylphenol, p-triphenylmethylphenol, phenol, 4-triphenylmethyl, p-cresol, .alpha.,.alpha.,.alpha.-triphenyl, 4-tritylphenol, p-triphenylmethyl phenol, 4-tri phenyl methyl phenol, phenol,4-triphenylmethyl
PubChem CID 70422
IUPAC Name 4-tritylphenol
SMILES OC1=CC=C(C=C1)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 276°C to 286°C
Quantity 5 g
Beilstein 2290531
Formula Weight 336.43
Percent Purity 98%
Chemical Name or Material 4-Tritylphenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 213-557-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only